N-Acryloxysuccinimide, 99%
N-Acryloxysuccinimide, 99%
N-Acryloxysuccinimide, 99%
N-Acryloxysuccinimide, 99%
Thermo Scientific Chemicals
N-Acryloxysuccinimide, 99%
CAS: 38862-24-7 | C7H7NO4 | 169.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA400300010
Price (MYR)
522.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
522.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Acryloxysuccinimide
CAS38862-24-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
View more
Coupling reagent
RUO – Research Use Only