D-Valine methyl ester hydrochloride, 99%
D-Valine methyl ester hydrochloride, 99%
D-Valine methyl ester hydrochloride, 99%
D-Valine methyl ester hydrochloride, 99%
Thermo Scientific Chemicals
D-Valine methyl ester hydrochloride, 99%
CAS: 7146-15-8 | C6H14ClNO2 | 167.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA398720050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Valine methyleester hydrochloride
CAS7146-15-8
ColorWhite
Infrared SpectrumAuthentic
Assay Percent Range99%