4-(Trifluoromethyl)styrene, 98+%, stabilized with 0.1% 4-tert-butylcatechol
CAS: 402-50-6 | C9H7F3 | 172.15 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FSA398460010
1 g
Glass bottle
Catalog number FSA398460010
Price (MYR)
637.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
637.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)styrene
Name NoteStabilized with 0.1% 4-tert-butylcatechol
CAS402-50-6
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning