Magnesium ethoxide, 97+%
Magnesium ethoxide, 97+%
Magnesium ethoxide, 97+%
Magnesium ethoxide, 97+%
Thermo Scientific Chemicals
Magnesium ethoxide, 97+%
CAS: 2414-98-4 | C4H10MgO2 | 114.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA397471000
Price (MYR)
605.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
605.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium ethoxide
CAS2414-98-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Self-heating; may catch fire.
Causes serious eye irritation.
Reacts violently with water.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
If