Cyclopentyl isocyanate, 97%
Cyclopentyl isocyanate, 97%
Cyclopentyl isocyanate, 97%
Cyclopentyl isocyanate, 97%
Thermo Scientific Chemicals
Cyclopentyl isocyanate, 97%
CAS: 4747-71-1 | C6H9NO | 111.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA397370050
Price (MYR)
891.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
891.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclopentyl isocyanate
CAS4747-71-1
Health Hazard 1GHS Signal Word: Danger
Density0.9860g/mL
Boiling Point147.0°C