6-Bromoindole, 96%
6-Bromoindole, 96%
6-Bromoindole, 96%
Thermo Scientific Chemicals
6-Bromoindole, 96%
CAS: 52415-29-9 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA397362500
Price (MYR)
382.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
382.00
EA
Add to cart
Specifications
Chemical Name or Material6-Bromoindole
CAS52415-29-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0