2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™
CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA397200010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyltetrahydrofuran
Melting Point-136.0°C
Name NoteStabilizer free, Extra Dry
CAS96-47-9
CAS Min %99.0