Check Order Status
7-Bromoindole, 98%, Thermo Scientific™
Element Search
Structure Search
7-Bromoindole, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number
FSA397062500
Price (MYR)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
7-Bromoindole
CAS
51417-51-7
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0