n-Octane, 99+%, Extra Dry, AcroSeal™
n-Octane, 99+%, Extra Dry, AcroSeal™
n-Octane, 99+%, Extra Dry, AcroSeal™
n-Octane, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
n-Octane, 99+%, Extra Dry, AcroSeal™
CAS: 111-65-9 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number FSA396900010
Price (MYR)
2,124.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
2,124.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Octane
Name NoteExtra Dry
CAS111-65-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger