Imidazole, ACS reagent
Imidazole, ACS reagent
Imidazole, ACS reagent
Imidazole, ACS reagent
Thermo Scientific Chemicals
Imidazole, ACS reagent
CAS: 288-32-4 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA396745000
Price (MYR)
846.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
846.00
EA
Add to cart
Specifications
Chemical Name or MaterialImidazole
Name NoteFor analysis ACS
CAS288-32-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May damage the unborn child.