1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals
1-Octanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
CAS: 5324-84-5 | C8H17NaO3S | 216.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA396690250
Price (MYR)
1,441.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,441.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octanesulfonic acid, sodium salt
Name NoteAnhydrous
CAS5324-84-5
Absorbance0.02 max. (C=0.4 M in water) at 260nm, 0.04 max. (C=0.4 M in water) at 230nm, 0.06 max. (C=0.4 M in water) at 220nm, 0.1 max. (C=0.4 M in water) at 210nm
CAS Min %99.0
View more
For ion pair chromatography
RUO – Research Use Only