1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA396671000
Price (MYR)
5,713.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,713.00
Each
Specifications
Chemical Name or Material1-Hexanesulfonic acid, sodium salt
Name NoteAnhydrous
CAS2832-45-3
Absorbance0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. (C=0.5 M in water) at 220nm, 0.1 max. (C=0.5 M in water) at 210nm
CAS Min %99.0
View more
For ion pair chromatography
RUO – Research Use Only