Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
Thermo Scientific Chemicals
Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA396400010
Price (MYR)
253.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
253.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340.0°C
CAS12125-02-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning