Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Thermo Scientific Chemicals
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
CAS: 5614-37-9 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA396132500
Price (MYR)
358.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
358.00
EA
Add to cart
Specifications
Chemical Name or MaterialCyclopentyl methyl ether
Name NoteStabilized
CAS5614-37-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May form explosive peroxides.