1-Butanol, for HPLC
1-Butanol, for HPLC
1-Butanol, for HPLC
Thermo Scientific Chemicals
1-Butanol, for HPLC
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA393750025
Price (MYR)
1,132.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,132.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butanol
Melting Point-89.0°C
Name NoteFor HPLC
Absorbance0.046 max. (1cm cell vs HLPC−grade water) at 245nm, 0.301 max. (1cm cell vs HPLC−grade water) at 220nm, 0.699 max. (1 cm cell vs HPLC−grade water) at 210nm
CAS71-36-3
View more
For HPLC
RUO – Research Use Only