3-Furaldehyde, 98%, stabilized
3-Furaldehyde, 98%, stabilized
3-Furaldehyde, 98%, stabilized
3-Furaldehyde, 98%, stabilized
Thermo Scientific Chemicals
3-Furaldehyde, 98%, stabilized
CAS: 498-60-2 | C5H4O2 | 96.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA393260050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Furaldehyde
CAS498-60-2
CAS Min %97.5
Health Hazard 1GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi
CAS Max %100.0