Ammonium bicarbonate, 99%, for analysis
Ammonium bicarbonate, 99%, for analysis
Ammonium bicarbonate, 99%, for analysis
Ammonium bicarbonate, 99%, for analysis
Thermo Scientific Chemicals
Ammonium bicarbonate, 99%, for analysis
CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA393212500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium bicarbonate
Melting Point105.0°C
CAS1066-33-7
ColorWhite
Health Hazard 1GHS Signal Word: Warning