Ammonium fluoride, 98+%, for analysis
Ammonium fluoride, 98+%, for analysis
Ammonium fluoride, 98+%, for analysis
Ammonium fluoride, 98+%, for analysis
Thermo Scientific Chemicals
Ammonium fluoride, 98+%, for analysis
CAS: 12125-01-8 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA393192500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium fluoride
CAS12125-01-8
ColorColorless to Yellow or White to Yellow
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.