3-Methoxybenzyl bromide, 98+%
3-Methoxybenzyl bromide, 98+%
3-Methoxybenzyl bromide, 98+%
3-Methoxybenzyl bromide, 98+%
Thermo Scientific Chemicals
3-Methoxybenzyl bromide, 98+%
CAS: 874-98-6 | C8H9BrO | 201.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA393110050
Price (MYR)
377.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
377.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Methoxybenzyl bromide
CAS874-98-6
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0