3-Iodobenzaldehyde, 99%
3-Iodobenzaldehyde, 99%
3-Iodobenzaldehyde, 99%
3-Iodobenzaldehyde, 99%
Thermo Scientific Chemicals
3-Iodobenzaldehyde, 99%
CAS: 696-41-3 | C7H5IO | 232.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA392070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Iodobenzaldehyde
CAS696-41-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0