4-Iodobenzaldehyde, 97+%
4-Iodobenzaldehyde, 97+%
4-Iodobenzaldehyde, 97+%
4-Iodobenzaldehyde, 97+%
Thermo Scientific Chemicals
4-Iodobenzaldehyde, 97+%
CAS: 15164-44-0 | C7H5IO | 232.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA392060010
Price (MYR)
726.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
726.00
EA
Add to cart
Specifications
Chemical Name or Material4-Iodobenzaldehyde
CAS15164-44-0
CAS Min %97.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0