(3-Chloropropyl)triethoxysilane, 97+%
(3-Chloropropyl)triethoxysilane, 97+%
(3-Chloropropyl)triethoxysilane, 97+%
(3-Chloropropyl)triethoxysilane, 97+%
Thermo Scientific Chemicals
(3-Chloropropyl)triethoxysilane, 97+%
CAS: 5089-70-3 | C9H21ClO3Si | 240.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA391890250
Price (MYR)
242.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
242.00
EA
Add to cart
Specifications
Chemical Name or Material(3-Chloropropyl)triethoxysilane
CAS5089-70-3
CAS Min %97.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0