2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
Thermo Scientific Chemicals
2-Iodobenzonitrile, 99%
CAS: 4387-36-4 | C7H4IN | 229.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA391800250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Iodobenzonitrile
CAS4387-36-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0