4-Chlorobenzyl bromide, 98%
4-Chlorobenzyl bromide, 98%
4-Chlorobenzyl bromide, 98%
4-Chlorobenzyl bromide, 98%
Thermo Scientific Chemicals
4-Chlorobenzyl bromide, 98%
CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA391710250
Price (MYR)
1,273.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,273.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chlorobenzyl bromide
CAS622-95-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0