Ammonium thiosulfate, 96%, pure
Ammonium thiosulfate, 96%, pure
Ammonium thiosulfate, 96%, pure
Ammonium thiosulfate, 96%, pure
Thermo Scientific Chemicals
Ammonium thiosulfate, 96%, pure
CAS: 7783-18-8 | H8N2O3S2 | 148.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA391665000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium thiosulfate
Melting Point150.0°C
Name NoteExtra Pure
CAS7783-18-8
ColorWhite
View more
Reduction reagent
RUO – Research Use Only