Titanium(IV) bromide, 98%
Titanium(IV) bromide, 98%
Titanium(IV) bromide, 98%
Titanium(IV) bromide, 98%
Thermo Scientific Chemicals
Titanium(IV) bromide, 98%
CAS: 7789-68-6 | TiBr4 | 367.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA391240250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point230.0°C
Chemical Name or MaterialTitanium(IV) bromide
Melting Point39.0°C
CAS7789-68-6
Health Hazard 1GHS Signal Word: Danger