Ammonium carbonate, extra pure
Ammonium carbonate, extra pure
Ammonium carbonate, extra pure
Ammonium carbonate, extra pure
Thermo Scientific Chemicals
Ammonium carbonate, extra pure
CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3902900101 kgPlastic bottle
Catalog number FSA390290010
Price (MYR)
338.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
338.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium carbonate
Melting Point58.0°C
CAS506-87-6
ColorWhite
Health Hazard 1GHS Signal Word: Warning