Isopropanol, 99.5+%, extra pure
Isopropanol, 99.5+%, extra pure
Isopropanol, 99.5+%, extra pure
Isopropanol, 99.5+%, extra pure
Thermo Scientific Chemicals
Isopropanol, 99.5+%, extra pure
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
Packaging:
Plastic Drum
Catalog number FSA389710100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
CAS67-63-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.