Potassium polysulfide, pure
Potassium polysulfide, pure
Potassium polysulfide, pure
Potassium polysulfide, pure
Thermo Scientific Chemicals
Potassium polysulfide, pure
CAS: 37199-66-9 | (S)nK2 | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA389592500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium polysulfide
Melting Point200.0°C to 250.0°C
CAS37199-66-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.