Activated charcoal, iodinated
Activated charcoal, iodinated
Activated charcoal, iodinated
Activated charcoal, iodinated
Activated charcoal, iodinated
CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA389470010
Price (MYR)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialActivated charcoal
CAS7440-44-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable solid.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
View more
Filter aid
RUO – Research Use Only