N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
CAS: 25561-30-2 | C8H18F3NOSi2 | 257.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA389325000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN, O-Bis(trimethylsilyl)trifluoroacetamide
CAS25561-30-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
CAS75-77-4
View more
Silylation reagent
RUO – Research Use Only