3-Mercapto-1-propanol, 97+%
3-Mercapto-1-propanol, 97+%
3-Mercapto-1-propanol, 97+%
3-Mercapto-1-propanol, 97+%
Thermo Scientific Chemicals
3-Mercapto-1-propanol, 97+%
CAS: 19721-22-3 | C3H8OS | 92.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA388570050
Price (MYR)
1,174.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,174.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Mercapto-1-propanol
CAS19721-22-3
CAS Min %97.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0