Poly(ethylene glycol methyl ether), average M.W. 500
Poly(ethylene glycol methyl ether), average M.W. 500
Poly(ethylene glycol methyl ether), average M.W. 500
Poly(ethylene glycol methyl ether), average M.W. 500
Thermo Scientific Chemicals
Poly(ethylene glycol methyl ether), average M.W. 500
CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA387890010
Price (MYR)
1,444.00
KG
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,444.00
KG
Add to cart
Specifications
Chemical Name or MaterialPoly(ethylene glycol methyl ether)
CAS9004-74-4
Melting Point12°C
ColorColorless
Density1.0800g/mL