3-bromo-n-methylaniline, 98%
3-bromo-n-methylaniline, 98%
3-bromo-n-methylaniline, 98%
3-bromo-n-methylaniline, 98%
Thermo Scientific Chemicals
3-bromo-n-methylaniline, 98%
CAS: 66584-32-5 | C7H8BrN | 186.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA387360050
Price (MYR)
1,210.00
G
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,210.00
G
Add to cart
Specifications
Chemical Name or Material3-Bromo-N-methylaniline
CAS66584-32-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0