3-Buten-2-ol, 97%
3-Buten-2-ol, 97%
3-Buten-2-ol, 97%
Thermo Scientific Chemicals
3-Buten-2-ol, 97%
CAS: 598-32-3 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA387070250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Buten-2-ol
CAS598-32-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0