View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIodine
CAS7553-56-2
ColorGray
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: Harmful if inhaled. Harmful in contact with skin. Very toxic to aquatic life.