(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
Thermo Scientific Chemicals
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA385331000
Price (MYR)
3,901.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,901.00
EA
Add to cart
Specifications
Chemical Name or Material(Trimethylsilyl)diazomethane
CAS18107-18-1
CAS Min %32.15
Health Hazard 1GHS Signal Word: Danger
CAS Max %35.54
View more
Alkylation reagent
RUO – Research Use Only