Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Titanium(IV) oxide, Aeroxide™ P25
Thermo Scientific Chemicals
Titanium(IV) oxide, Aeroxide™ P25
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA384292500
Price (MYR)
763.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
763.00
EA
Add to cart
Specifications
Boiling Point2900.0°C
Chemical Name or MaterialTitanium(IV) oxide
Melting Point1855.0°C
CAS13463-67-7
ColorWhite to Yellow
View more
Suitable for battery materials development.
RUO – Research Use Only