Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Isopropanol, 99.5%, for HPLC gradient grade
Thermo Scientific Chemicals
Isopropanol, 99.5%, for HPLC gradient grade
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Catalog number FSA383920025
Price (MYR)
1,120.00
LT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,120.00
LT
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Absorbance(1 cm cell vs HPLC grade water), (1 cm cell vs HPLC grade water) at 0nm, 0.005 max. at 260nm, 0.005 max. at 270nm, 0.01 max. at 250nm, 0.05 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, cut off at 205nm
CAS67-63-0
ColorColorless
View more
For HPLC
RUO – Research Use Only