3-Methyladenine, 90+%
3-Methyladenine, 90+%
3-Methyladenine, 90+%
3-Methyladenine, 90+%
Thermo Scientific Chemicals
3-Methyladenine, 90+%
CAS: 5142-23-4 | C6H7N5 | 149.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass Bottle
Catalog number FSA379791000
Price (MYR)
928.00
MG
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
928.00
MG
Add to cart
Specifications
Chemical Name or Material3-Methyladenine
CAS5142-23-4
CAS Min %90.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0