N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
N,N-Dimethylacetamide, 99.5%, extra pure
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99.5%, extra pure
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA378710025
Price (MYR)
807.00
LT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
807.00
LT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20.0°C
CAS127-19-5
CAS Min %99.4
Health Hazard 1Danger