Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Methyl-beta-cyclodextrin, average Mw 1310
Thermo Scientific Chemicals
Methyl-beta-cyclodextrin, average Mw 1310
CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA377110250
Price (MYR)
811.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
811.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl-β-cyclodextrin
Name NoteAverage Mn approx. 1310
CAS128446-36-6
Melting Point165.0°C to 172.0°C
ColorWhite
View more
Chelation/Complexation reagent
RUO – Research Use Only