4-Fluorophenylacetyl chloride, 98%
4-Fluorophenylacetyl chloride, 98%
4-Fluorophenylacetyl chloride, 98%
4-Fluorophenylacetyl chloride, 98%
Thermo Scientific Chemicals
4-Fluorophenylacetyl chloride, 98%
CAS: 459-04-1 | C8H6ClFO | 172.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA376120010
Price (MYR)
361.00
G
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
361.00
G
Add to cart
Specifications
Chemical Name or Material4-Fluorophenylacetyl chloride
CAS459-04-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0