Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Methyl acetate, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Methyl acetate, 99+%, Extra Dry, AcroSeal™
CAS: 79-20-9 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA371831000
Price (MYR)
357.00
ML
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
357.00
ML
Add to cart
Specifications
Chemical Name or MaterialMethyl acetate
CAS79-20-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.