L-Asparagine, 99%
L-Asparagine, 99%
L-Asparagine, 99%
Thermo Scientific Chemicals
L-Asparagine, 99%
CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number FSA371605000
Price (MYR)
1,119.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,119.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL-Asparagine
CAS70-47-3
ColorWhite
Infrared SpectrumAuthentic
Assay Percent Range99%