tert-Butyl acrylate, 99%, stabilized
tert-Butyl acrylate, 99%, stabilized
tert-Butyl acrylate, 99%, stabilized
tert-Butyl acrylate, 99%, stabilized
Thermo Scientific Chemicals
tert-Butyl acrylate, 99%, stabilized
CAS: 1663-39-4 | C7H12O2 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic drum
Catalog number FSA371130051
Price (MYR)
3,577.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
3,577.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butyl acrylate
Name Note99%
CAS1663-39-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.