Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Thermo Scientific Chemicals
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
CAS: 75-50-3 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA370785000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-50-3, 7732-18-5
Specifications
Chemical Name or MaterialTrimethylamine
ColorColorless
CAS75-50-3
Boiling Point30.0°C to 100.0°C
Health Hazard 1Danger