Eaton's reagent
Eaton's reagent
Eaton's reagent
Eaton's reagent
Thermo Scientific Chemicals
Eaton's reagent
CAS: 39394-84-8 | O10P4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Catalog number FSA369805000
Price (MYR)
1,024.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,024.00
BOT
Add to cart
Chemical Identifiers
CAS39394-84-8, 75-75-2, 1314-56-3
Specifications
Chemical Name or MaterialEaton's reagent
ColorUndesignated
CAS39394-84-8
Boiling Point122.0°C (1.0 mmHg)
Health Hazard 1GHS Signal Word: Danger