Palladium(II) chloride, 99.999%, (trace metal basis)
Palladium(II) chloride, 99.999%, (trace metal basis)
Palladium(II) chloride, 99.999%, (trace metal basis)
Palladium(II) chloride, 99.999%, (trace metal basis)
Thermo Scientific Chemicals
Palladium(II) chloride, 99.999%, (trace metal basis)
CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA369670250
Price (MYR)
31,922.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
31,922.00
EA
Add to cart
Specifications
Chemical Name or MaterialPalladium(II) Chloride
Melting Point678.0°C to 680.0°C
CAS7647-10-1
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
For ultra trace analysis
RUO – Research Use Only