View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4, 5-Dicyanoimidazole
Name Note99%
CAS1122-28-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger